Product Details

SNP ID
rs147779718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:12992895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGAGGCTCCCCATCCTCCAGGC[C/T]GCCTGCCAGCTCCCTGGACAGGAGG
Phenotype
MIM: 605367
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP44 PubMed Links

Gene Details

Gene
ARHGAP44
Gene Name
Rho GTPase activating protein 44
There are no transcripts associated with this gene.

Gene
ELAC2
Gene Name
elaC ribonuclease Z 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165962.1 2518 Missense Mutation AGC,GGC S762G NP_001159434.1
NM_018127.6 2518 Missense Mutation AGC,GGC S802G NP_060597.4
NM_173717.1 2518 Missense Mutation AGC,GGC S801G NP_776065.1

View Full Product Details