Product Details

SNP ID
rs148130302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81714994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGAGGTGAAGCTGCGCATGAC[A/G]GGCATGGCGCTGCGGGTGGTGCGTA
Phenotype
MIM: 602375 MIM: 606794
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL12 PubMed Links

Gene Details

Gene
MRPL12
Gene Name
mitochondrial ribosomal protein L12
There are no transcripts associated with this gene.

Gene
SLC25A10
Gene Name
solute carrier family 25 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270888.1 472 Silent Mutation ACA,ACG T45T NP_001257817.1
NM_001270953.1 472 Silent Mutation ACA,ACG T45T NP_001257882.1
NM_012140.4 472 Silent Mutation ACA,ACG T45T NP_036272.2
XM_017024220.1 472 Silent Mutation ACA,ACG T2T XP_016879709.1

View Full Product Details