Product Details

SNP ID
rs148192611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64547799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCTTTCCAAGTCCACAAATAAT[A/G]AGCTGTGAAAATAGTACACAGTAAT
Phenotype
MIM: 605532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP95 PubMed Links

Gene Details

Gene
CEP95
Gene Name
centrosomal protein 95
There are no transcripts associated with this gene.

Gene
SMURF2
Gene Name
SMAD specific E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022739.3 1860 Silent Mutation CTC,CTT L624L NP_073576.1
XM_005257585.3 1860 Silent Mutation CTC,CTT L611L XP_005257642.1
XM_011525128.1 1860 Silent Mutation CTC,CTT L626L XP_011523430.1

View Full Product Details