Product Details

SNP ID
rs148263909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44374468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGCCCGGAGCAGCTGTGTCCA[C/T]ACCTGGGGGCAAACCCACGTGTCTC
Phenotype
MIM: 607759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGA2B PubMed Links

Gene Details

Gene
ITGA2B
Gene Name
integrin subunit alpha 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000419.4 2876 Silent Mutation GTA,GTG V982V NP_000410.2
XM_011524749.1 2876 Silent Mutation GTA,GTG V948V XP_011523051.1
XM_011524750.1 2876 Intron XP_011523052.1

View Full Product Details