Product Details
- SNP ID
-
rs148265718
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:55720668 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCATTTGGTCGTATTCAGTCTC[A/G]AGCAAACAAGCTTCTCTGATTTGCC
- Phenotype
-
MIM: 616334
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM100
PubMed Links
Gene Details
- Gene
- TMEM100
- Gene Name
- transmembrane protein 100
There are no transcripts associated with this gene.
View Full Product Details