Product Details

SNP ID
rs148308911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41835931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCGCCCAGGCTGCCGCATCAG[A/G]ACCGTGGCCTTCATCAGGGTGCTCA
Phenotype
MIM: 608778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011524706.2 117 Intron XP_011523008.1
Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052935.4 117 Silent Mutation GTC,GTT V13V NP_443167.4
XM_006721669.3 117 UTR 5 XP_006721732.1
XM_006721670.3 117 UTR 5 XP_006721733.1
XM_011524276.2 117 Silent Mutation GTC,GTT V13V XP_011522578.1
XM_011524277.2 117 Silent Mutation GTC,GTT V5V XP_011522579.1
XM_017024127.1 117 Silent Mutation GTC,GTT V13V XP_016879616.1

View Full Product Details