Product Details

SNP ID
rs148311119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4785942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGACCCCTGCGCCCCACCTTCC[A/G]GACTGGGACTCTACCACGTGCGTAT
Phenotype
MIM: 604657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLTPD2 PubMed Links

Gene Details

Gene
GLTPD2
Gene Name
glycolipid transfer protein domain containing 2
There are no transcripts associated with this gene.

Gene
TM4SF5
Gene Name
transmembrane 4 L six family member 5
There are no transcripts associated with this gene.

Gene
VMO1
Gene Name
vitelline membrane outer layer 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144939.1 388 Silent Mutation TCC,TCT S102S NP_001138411.1
NM_001144940.1 388 Intron NP_001138412.1
NM_001144941.1 388 Intron NP_001138413.1
NM_182566.2 388 Silent Mutation TCC,TCT S102S NP_872372.1

View Full Product Details