Product Details

SNP ID
rs148323868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42663020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTGGGGGTACGGGTTCTGGCCT[A/G]GGCTCCTACCTCCTGGAGCGACTGA
Phenotype
MIM: 605785
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHH3 PubMed Links

Gene Details

Gene
PLEKHH3
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H3
There are no transcripts associated with this gene.

Gene
TUBG2
Gene Name
tubulin gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320509.1 703 Silent Mutation CTA,CTG L158L NP_001307438.1
NM_016437.2 703 Silent Mutation CTA,CTG L149L NP_057521.1
XM_011524614.2 703 Intron XP_011522916.1
XM_017024453.1 703 Intron XP_016879942.1
XM_017024454.1 703 UTR 5 XP_016879943.1
XM_017024455.1 703 UTR 5 XP_016879944.1
XM_017024456.1 703 UTR 5 XP_016879945.1
XM_017024457.1 703 UTR 5 XP_016879946.1

View Full Product Details