Product Details

SNP ID
rs148376521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42579540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCTCATACAAGATTTATTCCC[C/T]AGCACACCCCTCCCCTCCTCAGTTC
Phenotype
MIM: 616498 MIM: 602976 MIM: 608665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM134C PubMed Links

Gene Details

Gene
FAM134C
Gene Name
family with sequence similarity 134 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178126.3 3530 UTR 3 NP_835227.1
XM_011524440.2 3530 UTR 3 XP_011522742.1
Gene
MLX
Gene Name
MLX, MAX dimerization protein
There are no transcripts associated with this gene.

Gene
PSMC3IP
Gene Name
PSMC3 interacting protein
There are no transcripts associated with this gene.

View Full Product Details