Product Details
- SNP ID
-
rs148642752
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:1495859 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTGCTTCACCCAGTGGGTCCTC[C/T]CTCAAGGAGCCAGGCGGGATCTGCA
- Phenotype
-
MIM: 607875
MIM: 606538
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
INPP5K
PubMed Links
Gene Details
- Gene
- INPP5K
- Gene Name
- inositol polyphosphate-5-phosphatase K
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135642.1 |
1480 |
Silent Mutation |
AGA,AGG |
R361R |
NP_001129114.1 |
NM_016532.3 |
1480 |
Silent Mutation |
AGA,AGG |
R437R |
NP_057616.2 |
NM_130766.2 |
1480 |
Silent Mutation |
AGA,AGG |
R361R |
NP_570122.1 |
XM_005256683.2 |
1480 |
Silent Mutation |
AGA,AGG |
R361R |
XP_005256740.1 |
XM_005256685.1 |
1480 |
Silent Mutation |
AGA,AGG |
R345R |
XP_005256742.1 |
XM_005256686.1 |
1480 |
Silent Mutation |
AGA,AGG |
R345R |
XP_005256743.1 |
XM_011523934.1 |
1480 |
Silent Mutation |
AGA,AGG |
R361R |
XP_011522236.1 |
XM_011523936.2 |
1480 |
Silent Mutation |
AGA,AGG |
R302R |
XP_011522238.1 |
XM_017024755.1 |
1480 |
Silent Mutation |
AGA,AGG |
R361R |
XP_016880244.1 |
XM_017024756.1 |
1480 |
Silent Mutation |
AGA,AGG |
R361R |
XP_016880245.1 |
XM_017024757.1 |
1480 |
Silent Mutation |
AGA,AGG |
R345R |
XP_016880246.1 |
XM_017024758.1 |
1480 |
Silent Mutation |
AGA,AGG |
R302R |
XP_016880247.1 |
XM_017024759.1 |
1480 |
Silent Mutation |
AGA,AGG |
R302R |
XP_016880248.1 |
- Gene
- MYO1C
- Gene Name
- myosin IC
There are no transcripts associated with this gene.
View Full Product Details