Product Details

SNP ID
rs148850772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41026693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACTGTCCACAGTAGGACGGGC[A/G]GCAGCAGGAGGCCTGGGCATGGTGC
Phenotype
MIM: 608820 MIM: 608821 MIM: 608822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP1-3 PubMed Links

Gene Details

Gene
KRTAP1-3
Gene Name
keratin associated protein 1-3
There are no transcripts associated with this gene.

Gene
KRTAP1-4
Gene Name
keratin associated protein 1-4
There are no transcripts associated with this gene.

Gene
KRTAP1-5
Gene Name
keratin associated protein 1-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031957.1 510 Missense Mutation CGC,TGC R155C NP_114163.1

View Full Product Details