Product Details

SNP ID
rs148920521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42184169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAACGACTCAGATCCCGGACTG[C/T]CCTCCGGGCGCGACGGAGGCGGCGG
Phenotype
MIM: 608587 MIM: 602358 MIM: 604528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GHDC PubMed Links

Gene Details

Gene
GHDC
Gene Name
GH3 domain containing
There are no transcripts associated with this gene.

Gene
HCRT
Gene Name
hypocretin neuropeptide precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001524.1 468 Silent Mutation GGA,GGG G127G NP_001515.1
Gene
KCNH4
Gene Name
potassium voltage-gated channel subfamily H member 4
There are no transcripts associated with this gene.

View Full Product Details