Product Details

SNP ID
rs149012254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7442164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTATCCCACCTCCTCCAGATTTT[A/G]TCATTACTCACTTCCCTAGAGTCAT
Phenotype
MIM: 100710 MIM: 601514 MIM: 613936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNB1 PubMed Links

Gene Details

Gene
CHRNB1
Gene Name
cholinergic receptor nicotinic beta 1 subunit
There are no transcripts associated with this gene.

Gene
FGF11
Gene Name
fibroblast growth factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303460.1 Intron NP_001290389.1
NM_004112.3 Intron NP_004103.1
Gene
LOC107983988
Gene Name
proteoglycan 4-like
There are no transcripts associated with this gene.

Gene
TMEM102
Gene Name
transmembrane protein 102
There are no transcripts associated with this gene.

View Full Product Details