Product Details

SNP ID
rs149065653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48831824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGAGCTGAGTAAAATATGGATA[A/C]TATAACTTATTCTATTATTGGAAAC
Phenotype
MIM: 604587
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALCOCO2 PubMed Links

Gene Details

Gene
CALCOCO2
Gene Name
calcium binding and coiled-coil domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261390.1 Intron NP_001248319.1
NM_001261391.1 Intron NP_001248320.1
NM_001261393.1 Intron NP_001248322.1
NM_001261395.1 Intron NP_001248324.1
NM_005831.4 Intron NP_005822.1

View Full Product Details