Product Details

SNP ID
rs149067124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35122723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCTGTTCCTCCTCACCAAGTCT[G/T]GTTTTCCTCTACTTCATTCCACAAA
Phenotype
MIM: 602954
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FNDC8 PubMed Links

Gene Details

Gene
FNDC8
Gene Name
fibronectin type III domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017559.2 Intron NP_060029.1
Gene
NLE1
Gene Name
notchless homolog 1
There are no transcripts associated with this gene.

Gene
RAD51D
Gene Name
RAD51 paralog D
There are no transcripts associated with this gene.

Gene
RAD51L3-RFFL
Gene Name
RAD51L3-RFFL readthrough
There are no transcripts associated with this gene.

View Full Product Details