Product Details

SNP ID
rs149134550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41502687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGGCAGGAGGCTGCAGGCATAC[C/T]ATGCTCAGCTGGGACTGCAGCTCAA
Phenotype
MIM: 148065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT13 PubMed Links

Gene Details

Gene
KRT13
Gene Name
keratin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002274.3 1086 Missense Mutation ATA,ATG I341M NP_002265.2
NM_153490.2 1086 Missense Mutation ATA,ATG I341M NP_705694.2

View Full Product Details