Product Details

SNP ID
rs149137290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50479702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCTCGCCGCCTGGAGGAGGCTG[C/T]CATGCAGAAGTGTCTGGTGACCGAA
Phenotype
MIM: 610465 MIM: 602178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSF2 PubMed Links

Gene Details

Gene
ACSF2
Gene Name
acyl-CoA synthetase family member 2
There are no transcripts associated with this gene.

Gene
CHAD
Gene Name
chondroadherin
There are no transcripts associated with this gene.

Gene
RSAD1
Gene Name
radical S-adenosyl methionine domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018346.2 294 Missense Mutation GCC,GTC A70V NP_060816.1

View Full Product Details