Product Details

SNP ID
rs149210378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82658512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCCTTGCTCGGCCGCCAGAGCC[A/G]GTCCATCCCATGCCGCAGCAGCACG
Phenotype
MIM: 609226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4525 PubMed Links

Gene Details

Gene
MIR4525
Gene Name
microRNA 4525
There are no transcripts associated with this gene.

Gene
RAB40B
Gene Name
RAB40B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006822.2 782 Missense Mutation CGG,TGG R182W NP_006813.1
XM_005256334.4 782 Missense Mutation CGG,TGG R122W XP_005256391.1
XM_005256335.3 782 Intron XP_005256392.1
XM_006722271.3 782 Missense Mutation CGG,TGG R142W XP_006722334.1
XM_011523528.2 782 Missense Mutation CGG,TGG R165W XP_011521830.1
XM_017024042.1 782 Intron XP_016879531.1
XM_017024043.1 782 Missense Mutation CGG,TGG R122W XP_016879532.1
Gene
WDR45B
Gene Name
WD repeat domain 45B
There are no transcripts associated with this gene.

View Full Product Details