Product Details
- SNP ID
-
rs149321431
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41567625 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCCCCTGGACCCACTTCCTCCA[A/C]CATAGCCACCTCCACTTCCTCCTCC
- Phenotype
-
MIM: 607606
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3890472] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT9
- Gene Name
- keratin 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000226.3 |
1586 |
Missense Mutation |
GGT,GTT |
G507V |
NP_000217.2 |
View Full Product Details