Product Details

SNP ID
rs149344143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41378111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACAGGGTCCACAGGAGTTGCCA[C/G]TAGCATTGGTGGTGGCGCATGGGTT
Phenotype
MIM: 602762 MIM: 602763
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRT33B PubMed Links

Gene Details

Gene
KRT33B
Gene Name
keratin 33B
There are no transcripts associated with this gene.

Gene
KRT34
Gene Name
keratin 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021013.3 1350 Missense Mutation ACT,AGT T420S NP_066293.2
XM_011524793.1 1350 Missense Mutation ACT,AGT T396S XP_011523095.1

View Full Product Details