Product Details

SNP ID
rs149400522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60600870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTTTCTCGCTTGTCACCTTGC[C/T]ATGTGGAAGAAACTGGGTAAGTTCC
Phenotype
MIM: 605100
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPM1D PubMed Links

Gene Details

Gene
PPM1D
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003620.3 688 Silent Mutation GCC,GCT A152A NP_003611.1

View Full Product Details