Product Details

SNP ID
rs149443617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8174224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATCAGAAGGGTTTCTTTTTCAG[C/T]CCAAAGACGGTGACGAATAAAGCCA
Phenotype
MIM: 616663 MIM: 616183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNORD118 PubMed Links

Gene Details

Gene
SNORD118
Gene Name
small nucleolar RNA, C/D box 118
There are no transcripts associated with this gene.

Gene
TMEM107
Gene Name
transmembrane protein 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032354.3 512 Silent Mutation GGA,GGG G140G NP_115730.2
NM_183065.2 512 Silent Mutation GGA,GGG G134G NP_898888.1

View Full Product Details