Product Details

SNP ID
rs149468147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3610581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTAAGATTCCTTCTGGTTGAG[A/G]TGTCTCCGGAGCATGACCAGAGCTG
Phenotype
MIM: 605060 MIM: 602076
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHPK PubMed Links

Gene Details

Gene
SHPK
Gene Name
sedoheptulokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013276.2 1519 Silent Mutation CAC,CAT H472H NP_037408.2
Gene
TRPV1
Gene Name
transient receptor potential cation channel subfamily V member 1
There are no transcripts associated with this gene.

View Full Product Details