Product Details

SNP ID
rs149506389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48543667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTAAAGTGGAATTCCTTCTCCA[A/G]TTCCAGCAGCTGCGTGTTGGTGTAA
Phenotype
MIM: 142967 MIM: 142966
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXB-AS1 PubMed Links

Gene Details

Gene
HOXB-AS1
Gene Name
HOXB cluster antisense RNA 1
There are no transcripts associated with this gene.

Gene
HOXB2
Gene Name
homeobox B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002145.3 592 Silent Mutation CTG,TTG L158L NP_002136.1
XM_005257275.4 592 Silent Mutation CTG,TTG L49L XP_005257332.1
Gene
HOXB3
Gene Name
homeobox B3
There are no transcripts associated with this gene.

View Full Product Details