Product Details

SNP ID
rs149540824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21198837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGCAGTCTGTGGCCGACAAGCT[C/T]ACCCTTTTGATGGAGGGGGCAGGAA
Phenotype
MIM: 616160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS7B PubMed Links

Gene Details

Gene
DHRS7B
Gene Name
dehydrogenase/reductase 7B
There are no transcripts associated with this gene.

Gene
TMEM11
Gene Name
transmembrane protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003876.2 509 Silent Mutation GTA,GTG V22V NP_003867.1

View Full Product Details