Product Details

SNP ID
rs149555930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58155512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTTTTCTCTCAGTCATGGCCT[A/G]TGACCGCTACATAGCCATCTCCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSX2P1 PubMed Links

Gene Details

Gene
MSX2P1
Gene Name
msh homeobox 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
OR4D1
Gene Name
olfactory receptor family 4 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012374.1 359 Missense Mutation TAT,TGT Y120C NP_036506.1

View Full Product Details