Product Details

SNP ID
rs149700886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5420219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCGCGAAGAGGAGATGGCGGA[C/G]TCGTTGAGGTCTCCGCGCCGCTCCC
Phenotype
MIM: 602552
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUP88 PubMed Links

Gene Details

Gene
NUP88
Gene Name
nucleoporin 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320653.1 579 Intron NP_001307582.1
NM_002532.5 579 Intron NP_002523.2
XM_011523893.2 579 Intron XP_011522195.1
XM_017024694.1 579 Intron XP_016880183.1
Gene
RPAIN
Gene Name
RPA interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033002.3 579 Silent Mutation GAC,GAG D3E NP_001028174.2
NM_001160243.1 579 Silent Mutation GAC,GAG D3E NP_001153715.1
NM_001160244.1 579 Silent Mutation GAC,GAG D3E NP_001153716.1
NM_001160246.1 579 Silent Mutation GAC,GAG D3E NP_001153718.1
NM_001160266.1 579 Silent Mutation GAC,GAG D3E NP_001153738.1

View Full Product Details