Product Details

SNP ID
rs149804750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40477887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCTTATGGTGGGGCTCCCTGG[A/G]GAAGGAGATGCCTCTCCCGTCAGAC
Phenotype
MIM: 608385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNS4 PubMed Links

Gene Details

Gene
TNS4
Gene Name
tensin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032865.5 Intron NP_116254.4
XM_005257744.1 Intron XP_005257801.1
XM_017025236.1 Intron XP_016880725.1
XM_017025237.1 Intron XP_016880726.1

View Full Product Details