Product Details

SNP ID
rs149948770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82419877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTCCACCAAAGGTCTCGTACCT[A/C]TCAGAGGTAAGGACTCCCTTTTACC
Phenotype
MIM: 616864
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HEXDC PubMed Links

Gene Details

Gene
HEXDC
Gene Name
hexosaminidase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173620.2 192 Silent Mutation CTA,CTC L26L NP_775891.2
XM_005256346.3 192 UTR 5 XP_005256403.1
XM_005256347.4 192 Silent Mutation CTA,CTC L26L XP_005256404.1
XM_006722280.3 192 UTR 5 XP_006722343.1
XM_011523560.2 192 Intron XP_011521862.1
XM_011523561.2 192 Intron XP_011521863.1
XM_017024484.1 192 Silent Mutation CTA,CTC L26L XP_016879973.1
XM_017024485.1 192 UTR 5 XP_016879974.1
Gene
OGFOD3
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 3
There are no transcripts associated with this gene.

View Full Product Details