Product Details

SNP ID
rs150055368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41724795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGCATCTTCCACCTCAGCTGC[C/T]GCCTGTAATGGGCATCTTGCCAGTT
Phenotype
MIM: 137250 MIM: 600947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAST PubMed Links

Gene Details

Gene
GAST
Gene Name
gastrin
There are no transcripts associated with this gene.

Gene
HAP1
Gene Name
huntingtin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079870.1 1703 Missense Mutation CAG,CGG Q572R NP_001073339.1
NM_001079871.1 1703 Missense Mutation CAG,CGG Q564R NP_001073340.1
NM_177977.2 1703 Missense Mutation CAG,CGG Q589R NP_817084.2

View Full Product Details