Product Details

SNP ID
rs150213892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58347550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGACTCGCTGCCACTTGGAGA[A/C]CCAGCTGTCCTCTGGACTCATCATC
Phenotype
MIM: 612482 MIM: 603555
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF43 PubMed Links

Gene Details

Gene
RNF43
Gene Name
ring finger protein 43
There are no transcripts associated with this gene.

Gene
SUPT4H1
Gene Name
SPT4 homolog, DSIF elongation factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003168.2 Intron NP_003159.1
Gene
TSPOAP1-AS1
Gene Name
TSPOAP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details