Product Details

SNP ID
rs150448633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43399767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTGACTGAGATGGCGCCCACTG[C/G]CTCCTCCTTCTTGCCCCACTTCCAA
Phenotype
MIM: 600732
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL4D PubMed Links

Gene Details

Gene
ARL4D
Gene Name
ADP ribosylation factor like GTPase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001661.3 144 Missense Mutation GCC,GGC A12G NP_001652.2
XM_011524782.2 144 Missense Mutation GCC,GGC A12G XP_011523084.1

View Full Product Details