Product Details

SNP ID
rs150511114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80418532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGTCGGTATTCATTGTTCCGGCT[A/G]TGAAAGCTTAGACTAGGAAAGGATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENDOV PubMed Links

Gene Details

Gene
ENDOV
Gene Name
endonuclease V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164637.2 Intron NP_001158109.1
NM_001164638.1 Intron NP_001158110.1
NM_173627.4 Intron NP_775898.2
XM_005257244.3 Intron XP_005257301.1
XM_005257246.3 Intron XP_005257303.1
XM_006721837.3 Intron XP_006721900.1
XM_011524655.1 Intron XP_011522957.1
XM_011524656.1 Intron XP_011522958.1
XM_011524657.1 Intron XP_011522959.1
XM_011524658.2 Intron XP_011522960.1
XM_011524660.1 Intron XP_011522962.1
XM_011524661.2 Intron XP_011522963.1
XM_011524662.1 Intron XP_011522964.1
XM_011524663.1 Intron XP_011522965.1
XM_011524664.2 Intron XP_011522966.1
XM_011524666.1 Intron XP_011522968.1
XM_011524667.2 Intron XP_011522969.1
XM_011524669.2 Intron XP_011522971.1
XM_011524670.1 Intron XP_011522972.1
XM_011524671.2 Intron XP_011522973.1
XM_011524672.1 Intron XP_011522974.1
XM_011524673.1 Intron XP_011522975.1
XM_011524674.1 Intron XP_011522976.1
XM_011524675.1 Intron XP_011522977.1
XM_011524676.1 Intron XP_011522978.1
XM_011524677.1 Intron XP_011522979.1
XM_011524678.2 Intron XP_011522980.1
XM_017024508.1 Intron XP_016879997.1
XM_017024509.1 Intron XP_016879998.1
XM_017024510.1 Intron XP_016879999.1
XM_017024511.1 Intron XP_016880000.1
XM_017024512.1 Intron XP_016880001.1
XM_017024513.1 Intron XP_016880002.1
XM_017024514.1 Intron XP_016880003.1
XM_017024515.1 Intron XP_016880004.1
XM_017024516.1 Intron XP_016880005.1
XM_017024517.1 Intron XP_016880006.1
XM_017024518.1 Intron XP_016880007.1
XM_017024519.1 Intron XP_016880008.1
XM_017024520.1 Intron XP_016880009.1
Gene
LOC100294362
Gene Name
uncharacterized LOC100294362
There are no transcripts associated with this gene.

Gene
MIR4730
Gene Name
microRNA 4730
There are no transcripts associated with this gene.

View Full Product Details