Product Details

SNP ID
rs150545240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81834585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCTCGCTGGTGAATGTCAGCCC[C/G]GTATTTGACCAGCAGCTTCACGCAT
Phenotype
MIM: 616514 MIM: 176790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MCRIP1 PubMed Links

Gene Details

Gene
MCRIP1
Gene Name
MAPK regulated corepressor interacting protein 1
There are no transcripts associated with this gene.

Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
There are no transcripts associated with this gene.

Gene
PPP1R27
Gene Name
protein phosphatase 1 regulatory subunit 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007533.3 356 Missense Mutation CGG,GGG R87G NP_001007534.1

View Full Product Details