Product Details

SNP ID
rs150552930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28714689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTGCTGGCAGTTAGGTAGAGG[C/T]TGCTGTCATCACTGTTGATGCCTAA
Phenotype
MIM: 610917 MIM: 602326
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NARR PubMed Links

Gene Details

Gene
NARR
Gene Name
nine-amino acid residue-repeats
There are no transcripts associated with this gene.

Gene
PROCA1
Gene Name
protein interacting with cyclin A1
There are no transcripts associated with this gene.

Gene
RAB34
Gene Name
RAB34, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142624.2 906 Missense Mutation AAC,AGC N294S NP_001136096.2
NM_001142625.2 906 Intron NP_001136097.2
NM_001144942.1 906 Missense Mutation AAC,AGC N237S NP_001138414.1
NM_001144943.1 906 Missense Mutation AAC,AGC N302S NP_001138415.1
NM_001256276.1 906 Missense Mutation AAC,AGC N223S NP_001243205.1
NM_001256277.1 906 Missense Mutation AAC,AGC N245S NP_001243206.1
NM_001256278.1 906 Missense Mutation AAC,AGC N245S NP_001243207.1
NM_031934.5 906 Missense Mutation AAC,AGC N245S NP_114140.4
Gene
RPL23A
Gene Name
ribosomal protein L23a
There are no transcripts associated with this gene.

Gene
SNORD42A
Gene Name
small nucleolar RNA, C/D box 42A
There are no transcripts associated with this gene.

Gene
SNORD42B
Gene Name
small nucleolar RNA, C/D box 42B
There are no transcripts associated with this gene.

Gene
SNORD4A
Gene Name
small nucleolar RNA, C/D box 4A
There are no transcripts associated with this gene.

Gene
SNORD4B
Gene Name
small nucleolar RNA, C/D box 4B
There are no transcripts associated with this gene.

Gene
TLCD1
Gene Name
TLC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details