Product Details

SNP ID
rs150652834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82246015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGGGAGACGAGCAGCTACTTGC[C/T]GTGGTGTTCGAAAGGAATGCTATTC
Phenotype
MIM: 600864 MIM: 603877
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSNK1D PubMed Links

Gene Details

Gene
CSNK1D
Gene Name
casein kinase 1 delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001893.4 1559 Intron NP_001884.2
NM_139062.2 1559 Missense Mutation AGC,GGC S408G NP_620693.1
XM_005256336.3 1559 Intron XP_005256393.1
XM_005256337.4 1559 Intron XP_005256394.1
XM_017024199.1 1559 Intron XP_016879688.1
Gene
MIR6787
Gene Name
microRNA 6787
There are no transcripts associated with this gene.

Gene
SLC16A3
Gene Name
solute carrier family 16 member 3
There are no transcripts associated with this gene.

View Full Product Details