Product Details

SNP ID
rs150690901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61401918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGTGGCCTTCCACAAGCTGAA[A/G]CTGACCAACAACATCTCTGACAAGC
Phenotype
MIM: 607470 MIM: 600747
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAS3 PubMed Links

Gene Details

Gene
BCAS3
Gene Name
BCAS3, microtubule associated cell migration factor
There are no transcripts associated with this gene.

Gene
C17orf82
Gene Name
chromosome 17 open reading frame 82
There are no transcripts associated with this gene.

Gene
TBX2
Gene Name
T-box 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005994.3 911 Silent Mutation AAA,AAG K210K NP_005985.3
XM_011525159.1 911 Intron XP_011523461.1
Gene
TBX2-AS1
Gene Name
TBX2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details