Product Details

SNP ID
rs150738879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41379698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGTCCACCTCCACGTTGAGGC[A/G]GTCTCCAAGCTGGGAGCGCAGGGTG
Phenotype
MIM: 602762 MIM: 602763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT33B PubMed Links

Gene Details

Gene
KRT33B
Gene Name
keratin 33B
There are no transcripts associated with this gene.

Gene
KRT34
Gene Name
keratin 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021013.3 839 Missense Mutation CGC,TGC R250C NP_066293.2
XM_011524793.1 839 Missense Mutation CGC,TGC R226C XP_011523095.1

View Full Product Details