Product Details

SNP ID
rs150742579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49973251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGCCCAGCTGGCAGCGCAGCT[A/C]GGCCTCACCCAGACCCAGGTGGGGC
Phenotype
MIM: 601911
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DLX4 PubMed Links

Gene Details

Gene
DLX4
Gene Name
distal-less homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001934.3 779 Silent Mutation CTA,CTC L82L NP_001925.2
NM_138281.2 779 Silent Mutation CTA,CTC L154L NP_612138.1
XM_017024291.1 779 Silent Mutation CTA,CTC L82L XP_016879780.1

View Full Product Details