Product Details

SNP ID
rs151076113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7260677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGATCAAAGGGTTATAAAAGTC[C/T]GTGACAATCTGATGGCCATACCAGG
Phenotype
MIM: 609131 MIM: 610684 MIM: 615019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN7 PubMed Links

Gene Details

Gene
CLDN7
Gene Name
claudin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185022.1 1340 Silent Mutation ACA,ACG T146T NP_001171951.1
NM_001185023.1 1340 Intron NP_001171952.1
NM_001307.5 1340 Silent Mutation ACA,ACG T146T NP_001298.3
Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
There are no transcripts associated with this gene.

Gene
ELP5
Gene Name
elongator acetyltransferase complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015362.4 1340 Intron NP_056177.3
NM_203413.2 1340 Intron NP_981958.1
NM_203414.2 1340 Intron NP_981959.1
NM_203415.1 1340 Intron NP_981960.1
XM_011523779.2 1340 Intron XP_011522081.1

View Full Product Details