Product Details

SNP ID
rs151173660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40313874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGAGAAATCAATAGTGCAAAT[G/T]GATTGAGGCTGTTTAGCCTGTCAAC
Phenotype
MIM: 180240
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371934 PubMed Links

Gene Details

Gene
LOC105371934
Gene Name
uncharacterized LOC105371934
There are no transcripts associated with this gene.

Gene
RARA
Gene Name
retinoic acid receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000964.3 Intron NP_000955.1
NM_001024809.3 Intron NP_001019980.1
NM_001145301.2 Intron NP_001138773.1
NM_001145302.2 Intron NP_001138774.1
XM_005257552.4 Intron XP_005257609.1
XM_005257553.1 Intron XP_005257610.1
XM_005257554.1 Intron XP_005257611.1
XM_011525095.1 Intron XP_011523397.1
XM_011525096.1 Intron XP_011523398.1
XM_017024920.1 Intron XP_016880409.1

View Full Product Details