Product Details

SNP ID
rs151252462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35735302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTGCGTCCCCACCACCGCCCGC[C/T]GCCTTTACCTGCCTGCTGTCGTCAT
Phenotype
MIM: 601025 MIM: 611398 MIM: 612128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP2B1 PubMed Links

Gene Details

Gene
AP2B1
Gene Name
adaptor related protein complex 2 beta 1 subunit
There are no transcripts associated with this gene.

Gene
GAS2L2
Gene Name
growth arrest specific 2 like 2
There are no transcripts associated with this gene.

Gene
RASL10B
Gene Name
RAS like family 10 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033315.3 484 Missense Mutation CGC,TGC R40C NP_201572.1
XM_017025300.1 484 Missense Mutation CGC,TGC R99C XP_016880789.1
XM_017025301.1 484 Missense Mutation CGC,TGC R40C XP_016880790.1

View Full Product Details