Product Details

SNP ID
rs151256408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76081661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCTCCGGGCCATTGAGCGCAT[A/G]GGCTACAAGGTGACATTGCTGCTGA
Phenotype
MIM: 608163 MIM: 603691 MIM: 604858 MIM: 610935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC7 PubMed Links

Gene Details

Gene
EXOC7
Gene Name
exocyst complex component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013839.3 869 UTR 3 NP_001013861.1
NM_001145297.3 869 UTR 3 NP_001138769.1
NM_001145298.3 869 UTR 3 NP_001138770.1
NM_001145299.3 869 UTR 3 NP_001138771.1
NM_001282313.1 869 UTR 3 NP_001269242.1
NM_001282314.1 869 Intron NP_001269243.1
NM_015219.4 869 UTR 3 NP_056034.2
XM_006721786.3 869 UTR 3 XP_006721849.1
XM_006721787.3 869 UTR 3 XP_006721850.1
XM_006721788.3 869 UTR 3 XP_006721851.1
XM_006721789.3 869 UTR 3 XP_006721852.1
XM_011524571.2 869 UTR 3 XP_011522873.1
XM_011524572.2 869 Intron XP_011522874.1
XM_017024397.1 869 UTR 3 XP_016879886.1
Gene
GALR2
Gene Name
galanin receptor 2
There are no transcripts associated with this gene.

Gene
SRP68
Gene Name
signal recognition particle 68
There are no transcripts associated with this gene.

Gene
ZACN
Gene Name
zinc activated ion channel
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_180990.3 869 Missense Mutation ATA,ATG I262M NP_851321.2

View Full Product Details