Product Details

SNP ID
rs151272358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82374627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGCTGGCCGACCTGCTGTACCT[A/G]CTCAGCATCCCCTTCATCGTGGCCA
Phenotype
MIM: 600896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376790 PubMed Links

Gene Details

Gene
LOC105376790
Gene Name
uncharacterized LOC105376790
There are no transcripts associated with this gene.

Gene
UTS2R
Gene Name
urotensin 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018949.2 351 Silent Mutation CTA,CTG L101L NP_061822.1

View Full Product Details