Product Details

SNP ID
rs151293476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74924835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGACGTGTTCGCGCTGCTCACTG[C/T]GATGATGCTGCTGGCAACGCTCTGG
Phenotype
MIM: 607827 MIM: 607696
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178160.2 295 Missense Mutation GCG,GTG A68V NP_835454.1
XM_011525479.1 295 Missense Mutation GCG,GTG A68V XP_011523781.1
Gene
USH1G
Gene Name
USH1 protein network component sans
There are no transcripts associated with this gene.

View Full Product Details