Product Details

SNP ID
rs138304922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:321787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATGGCCATGACCCCAGTTATCC[A/G]GCTGTCCAGCTTTCCAATTTCTTTT
Phenotype
MIM: 607621
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COLEC12 PubMed Links

Gene Details

Gene
COLEC12
Gene Name
collectin subfamily member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130386.2 2157 Missense Mutation CCG,CTG P695L NP_569057.1
XM_011525741.2 2157 Missense Mutation CCG,CTG P678L XP_011524043.1

View Full Product Details