Product Details

SNP ID
rs140095500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54155225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGCTCTATGTGCTCGGGTACA[C/T]TTTTTTTCTTACAGGCAAAAGCCAG
Phenotype
MIM: 603547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBD2 PubMed Links

Gene Details

Gene
MBD2
Gene Name
methyl-CpG binding domain protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003927.4 1564 UTR 3 NP_003918.1
NM_015832.4 1564 Intron NP_056647.1

View Full Product Details