Product Details

SNP ID
rs140107443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63393489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGGCACCAGGGTCACCTGGAG[A/G]GCAAGGTGTTAGCAGATCATCTACA
Phenotype
MIM: 609983
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS4B PubMed Links

Gene Details

Gene
VPS4B
Gene Name
vacuolar protein sorting 4 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004869.3 1413 Missense Mutation CCT,TCT P385S NP_004860.2
XM_006722582.2 1413 Missense Mutation CCT,TCT P267S XP_006722645.1
XM_011526268.2 1413 Missense Mutation CCT,TCT P385S XP_011524570.1

View Full Product Details