Product Details

SNP ID
rs141065471
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:77250996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTCCTCCTCCCTCAGGGTGTCC[C/G]GCAACGCGCTGCTGGGCGTGGGCTG
Phenotype
MIM: 600377
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GALR1 PubMed Links

Gene Details

Gene
GALR1
Gene Name
galanin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001480.3 945 Missense Mutation CGC,GGC R150G NP_001471.2
XM_017025691.1 945 Missense Mutation CGC,GGC R150G XP_016881180.1

View Full Product Details