Product Details

SNP ID
rs141535428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23463026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGCTGATGTAATGAGTGAACA[A/G]CTGGCCAAAGAATTGCAGTTAGAAG
Phenotype
MIM: 603579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIOK3 PubMed Links

Gene Details

Gene
RIOK3
Gene Name
RIO kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003831.3 743 Silent Mutation CAA,CAG Q42Q NP_003822.2
XM_011526242.1 743 UTR 5 XP_011524544.1
XM_011526243.1 743 UTR 5 XP_011524545.1

View Full Product Details